Skip to Content
DK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
DL-α-Tocopherol acetate
Synonym(s):
Vitamin E acetate, all-
rac
-α-Tocopheryl acetate
Empirical Formula (Hill Notation):
C
31
H
52
O
3
CAS Number:
7695-91-2
Molecular Weight:
472.74
EC Number:
231-710-0
Products
Product Category
vitamins (2)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T3376
DL-α-Tocopherol acetate,
≥96% (HPLC)
Expand
View Pricing
29992
DL-α-Tocopherol acetate,
tested according to Ph. Eur.
Expand
View Pricing