Silver nitrate
Synonym(s):
Nitric acid silver(I) salt, Silver nitrate
Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
Products
Product Category
Brand
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing