Skip to Content
Merck
Dextran from Leuconostoc mesenteroides average mol wt 150,000

Dextran from Leuconostoc spp.

Dextran from Leuconostoc mesenteroides average mol wt 150,000
Linear Formula:
(C6H10O5)n
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Manufacturer
Physical Form
Format
Analyte Suitability
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-20 of 55
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3