Molecular sieve 0.3 nm
Synonym(s):
Molecular sieve 0.3 nm, Molecular sieve 0.4 nm, Molecular sieve 1.0 nm
Linear Formula:
(SiO2)x(Al2O3)y
CAS Number:
Products
Product Category
Brand
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Particle Size (µm)
Technique