Skip to Content
DK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tryptone (vegetable)
Synonym(s):
Peptone from vegetable
CAS Number:
100209-45-8
EC Number:
309-353-8
Products
Product Category
peptones (11)
Brand
Millipore (9)
Biological Source
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
16922
Tryptone (vegetable),
suitable for microbiology, meat-free alternative
Expand
View Pricing
83059
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
19942
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
29185
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
18332
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
61854
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
38143
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
96174
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
92976
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
51841
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing
07436
Tryptone (vegetable),
suitable for microbiology
Expand
View Pricing