Triethylamine
Synonym(s):
N,N-Diethylethanamine, Triethylamine
Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing