- Zfp462 deficiency causes anxiety-like behaviors with excessive self-grooming in mice.
Zfp462 deficiency causes anxiety-like behaviors with excessive self-grooming in mice.
Genes, brain, and behavior (2016-09-14)
B Wang, Y Zheng, H Shi, X Du, Y Zhang, B Wei, M Luo, H Wang, X Wu, X Hua, M Sun, X Xu
PMID27621227
ABSTRACT
Zfp462 is a newly identified vertebrate-specific zinc finger protein that contains nearly 2500 amino acids and 23 putative C2H2-type zinc finger domains. So far, the functions of Zfp462 remain unclear. In our study, we showed that Zfp462 is expressed predominantly in the developing brain, especially in the cerebral cortex and hippocampus regions from embryonic day 7.5 to early postnatal stage. By using a piggyBac transposon-generated Zfp462 knockout (KO) mouse model, we found that Zfp462 KO mice exhibited prenatal lethality with normal neural tube patterning, whereas heterozygous (Het) Zfp462 KO (Zfp462