Merck
  • Distinct expression and function of alternatively spliced Tbx5 isoforms in cell growth and differentiation.
Distinct expression and function of alternatively spliced Tbx5 isoforms in cell growth and differentiation.
Molecular and cellular biology (2008-04-09)
Romain Georges, Georges Nemer, Martin Morin, Chantal Lefebvre, Mona Nemer
ABSTRACT
Mutations in the T-box transcription factor Tbx5 cause Holt-Oram syndrome, an autosomal dominant disease characterized by a wide spectrum of cardiac and upper limb defects with variable expressivity. Tbx5 haploinsufficiency has been suggested to be the underlying mechanism, and experimental models are consistent with a dosage-sensitive requirement for Tbx5 in heart development. Here, we report that Tbx5 levels are regulated through alternative splicing that generates, in addition to the known 518-amino-acid protein, a C-terminal truncated isoform. This shorter isoform retains the capacity to bind DNA, but its interaction with Tbx5 collaborators such as GATA-4 is altered. In vivo, the two spliced isoforms are oppositely regulated in a temporal and growth factor-dependent manner and are present in distinct DNA-binding complexes. The expression of the long isoform correlates with growth stimulation, and its reexpression in postnatal transgenic mouse hearts promotes hypertrophy. Conversely, the upregulation of the short but not the long isoform in C2C12 myoblasts leads to growth arrest and cell death. The results provide novel insight into posttranscriptional Tbx5 regulation and point to an important role not only in cell differentiation but also in cell proliferation and organ growth. The data may help analyze genotype-phenotype relations in patients with Holt-Oram syndrome.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M5 antibody produced in mouse, clone M5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)