Merck
  • Involvement of VAT-1 in Phosphatidylserine Transfer from the Endoplasmic Reticulum to Mitochondria.
Involvement of VAT-1 in Phosphatidylserine Transfer from the Endoplasmic Reticulum to Mitochondria.
Traffic (Copenhagen, Denmark) (2015-09-24)
Mirco Junker, Tom A Rapoport
ABSTRACT
Mitochondria receive phosphatidylserine (PS) from the endoplasmic reticulum (ER), but how PS is moved from the ER to mitochondria is unclear. Current models postulate a physical link between the organelles, but no involvement of cytosolic proteins. Here, we have reconstituted PS transport from the ER to mitochondria in vitro using Xenopus egg components. Transport is independent of ER proteins, but is dependent on a cytosolic factor that has a preferential affinity for PS. Crosslinking with a photoactivatable PS analog identified VAT-1 as a candidate for a cytosolic PS transport protein. Recombinant, purified VAT-1 stimulated PS transport into mitochondria and depletion of VAT-1 from Xenopus cytosol with specific antibodies led to a reduction of transport. Our results suggest that cytosolic factors have a role in PS transport from the ER to mitochondria, implicate VAT-1 in the transport process, and indicate that physical contact between the organelles is not essential.
Sigma-Aldrich
Tris(3-hydroxypropyltriazolylmethyl)amine, 95%
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder