- SNIP/p140Cap mRNA expression is an unfavourable prognostic factor in breast cancer and is not expressed in normal breast tissue.
SNIP/p140Cap mRNA expression is an unfavourable prognostic factor in breast cancer and is not expressed in normal breast tissue.
British journal of cancer (2008-05-14)
S Kennedy, M Clynes, P Doolan, J P Mehta, S Rani, J Crown, L O'Driscoll
PMID18475297
ABSTRACT
The prevalence and clinical relevance of SNIP/p140Cap has not been extensively investigated. Here SNIP/p140Cap mRNA expression was studied in 103 breast tumour biopsies, where it was detected in approximately 37% of tumour specimens, but not in any normal breast specimens. Expression correlated significantly with unfavourable overall survival. This suggests that SNIP/p140Cap may be a useful diagnostic and prognostic marker for breast cancer and its expression in breast cancer, but not in normal breast tissue, suggests that it may have potential as a therapeutic target.