Merck
  • Pioglitazone reduces angiotensin II-induced COX-2 expression through inhibition of ROS production and ET-1 transcription in vascular cells from spontaneously hypertensive rats.
Pioglitazone reduces angiotensin II-induced COX-2 expression through inhibition of ROS production and ET-1 transcription in vascular cells from spontaneously hypertensive rats.
American journal of physiology. Heart and circulatory physiology (2014-04-15)
Jose V Pérez-Girón, Roberto Palacios, Angela Martín, Raquel Hernanz, Andrea Aguado, Sonia Martínez-Revelles, María T Barrús, Mercedes Salaices, María J Alonso
ABSTRACT
Glitazones have anti-inflammatory properties by interfering with the transcription of proinflammatory genes, such as cyclooxygenase (COX)-2, and with ROS production, which are increased in hypertension. This study analyzed whether pioglitazone modulates COX-2 expression in hypertension by interfering with ROS and endothelin (ET)-1. In vivo, pioglitazone (2.5 mg·kg(-1)·day(-1), 28 days) reduced the greater levels of COX-2, pre-pro-ET-1, and NADPH oxidase (NOX) expression and activity as well as O2 (·-) production found in aortas from spontaneously hypertensive rats (SHRs). ANG II increased COX-2 and pre-pro-ET-1 levels more in cultured vascular smooth muscle cells from hypertensive rats compared with normotensive rats. The ETA receptor antagonist BQ-123 reduced ANG II-induced COX-2 expression in SHR cells. ANG II also increased NOX-1 expression, NOX activity, and superoxide production in SHR cells; the selective NOX-1 inhibitor ML-171 and catalase reduced ANG II-induced COX-2 and ET-1 transcription. ANG II also increased c-Jun transcription and phospho-JNK1/2, phospho-c-Jun, and p65 NF-κB subunit nuclear protein expression. SP-600125 and lactacystin, JNK and NF-κB inhibitors, respectively, reduced ANG II-induced ET-1, COX-2, and NOX-1 levels and NOX activity. Pioglitazone reduced the effects of ANG II on NOX activity, NOX-1, pre-pro-ET-1, COX-2, and c-Jun mRNA levels, JNK activation, and nuclear phospho-c-Jun and p65 expression. In conclusion, ROS production and ET-1 are involved in ANG II-induced COX-2 expression in SHRs, explaining the greater COX-2 expression observed in this strain. Furthermore, pioglitazone inhibits ANG II-induced COX-2 expression likely by interfering with NF-κB and activator protein-1 proinflammatory pathways and downregulating ROS production and ET-1 transcription, thus contributing to the anti-inflammatory properties of glitazones.
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%