Merck
  • Asymmetrically simultaneous synthesis of L-homophenylalanine and N6-protected-2-oxo-6-amino-hexanoic acid by engineered Escherichia coli aspartate aminotransferase.
Asymmetrically simultaneous synthesis of L-homophenylalanine and N6-protected-2-oxo-6-amino-hexanoic acid by engineered Escherichia coli aspartate aminotransferase.
Biotechnology progress (2009-09-22)
Hsueh-Hsia Lo, Wei-De Lin, Shih-Kuang Hsu, Wen-Hwei Hsu
ABSTRACT
L-Homophenylalanine (L-HPA) and N(6)-protected-2-oxo-6-amino-hexanoic acid (N(6)-protected-OAHA) can be used as building blocks for the manufacture of angiotensin-converting enzyme inhibitors. To synthesize L-HPA and N(6)-protected-OAHA simultaneously from 2-oxo-4-phenylbutanoic acid (OPBA) and N(6)-protected-L-lysine, several variants of Escherichia coli aspartate aminotransferase (AAT) were developed by site-directed mutagenesis and their catalytic activities were investigated. Three kinds of N(6)-protected-L-lysine were tested as potential amino donors for the bioconversion process. AAT variants of R292E/L18H and R292E/L18T exhibited specific activities of 0.70+/-0.01 U/mg protein and 0.67+/-0.02 U/mg protein to 2-amino-6-tert-butoxycarbonylamino-hexanoic acid (BOC-lysine) and 2-amino-6-(2,2,2-trifluoro-acetylamino)-hexanoic acid, respectively. E. coli cells expressing R292E/L18H variant were able to convert OPBA and BOC-lysine to L-HPA and 2-oxo-6-tert-butoxycarbonylamino-hexanoic acid (BOC-OAHA) with 96.2% yield in 8 h. This is the first report demonstrating a process for the simultaneous production of two useful building blocks, L-HPA and BOC-OAHA.
Sigma-Aldrich
L-Homophenylalanine hydrochloride, 97%
Sigma-Aldrich
DL-Homophenylalanine, 98%