Merck
  • Induced Polyspecificity of Human Secretory Immunoglobulin A Antibodies: Is It Possible to Improve Their Ability to Bind Pathogens?
Induced Polyspecificity of Human Secretory Immunoglobulin A Antibodies: Is It Possible to Improve Their Ability to Bind Pathogens?
Pharmacology (2021-12-06)
Ekaterina N Gorshkova, Shina Pashova, Ekaterina A Vasilenko, Tatiana S Tchurina, Elizaveta A Razzorenova, Olga V Starkina, Petya Dimitrova, Anastas Pashov, Tchavdar Lubenov Vassilev
ABSTRACT
As has been shown previously, various protein-modifying agents can change the antigen-binding properties of immunoglobulins. However, induced polyspecificity of human secretory immunoglobulin A (sIgA) has not been previously characterized in detail. In the present study, human secretory immunoglobulin A (IgA) was exposed to buffers with acidic pH, to free heme, or to pro-oxidative ferrous ions, and the antigen-binding behavior of the native and modified IgA to viral and bacterial antigens was compared using Western blotting and enzyme-linked immunosorbent assay. The ability of these agents to modulate the antigen-binding properties of human sIgA toward a wide range of pathogen peptides was investigated using an epitope microarray. We have shown that acidic pH, heme, and pro-oxidative ferrous ions influenced the binding of secretory IgA in opposite directions (either increasing or decreasing); however, the strongest effect was observed when using buffers with low pH. This fraction had the highest number of affected reactivities; most of them were increased and most of the new ones were toward common pathogens. Thus, it was shown that all investigated treatments can alter to some degree the antigen-binding of secretory IgA, but acidic pH has the most potentially beneficial effect by increasing binding to a largest number of common pathogens' antigens.
Sigma-Aldrich
IgA from human colostrum, reagent grade, buffered aqueous solution