Merck
  • Host dysbiosis negatively impacts IL-9-producing T-cell differentiation and antitumour immunity.
Host dysbiosis negatively impacts IL-9-producing T-cell differentiation and antitumour immunity.
British journal of cancer (2020-06-06)
Rafael Ribeiro Almeida, Raquel de Souza Vieira, Angela Castoldi, Fernanda Fernandes Terra, Amanda Campelo L Melo, Maria Cecília Campos Canesso, Luísa Lemos, Marcella Cipelli, Nisha Rana, Meire Ioshie Hiyane, Erika L Pearce, Flaviano Dos Santos Martins, Ana Maria Caetano de Faria, Niels Olsen Saraiva Câmara
ABSTRACT
Host-microbiota interactions shape T-cell differentiation and promote tumour immunity. Although IL-9-producing T cells have been described as potent antitumour effectors, their role in microbiota-mediated tumour control remains unclear. We analysed the impact of the intestinal microbiota on the differentiation of colonic lamina propria IL-9-producing T cells in germ-free and dysbiotic mice. Systemic effects of the intestinal microbiota on IL-9-producing T cells and the antitumour role of IL-9 were analysed in a model of melanoma-challenged dysbiotic mice. We show that germ-free mice have lower frequency of colonic lamina propria IL-9-producing T cells when compared with conventional mice, and that intestinal microbiota reconstitution restores cell frequencies. Long-term antibiotic treatment promotes host dysbiosis, diminishes intestinal IL-4 and TGF-β gene expression, decreases the frequency of colonic lamina propria IL-9-producing T cells, increases the susceptibility to tumour development and reduces the frequency of IL-9-producing T cells in the tumour microenvironment. Faecal transplant restores intestinal microbiota diversity, and the frequency of IL-9-producing T cells in the lungs of dysbiotic animals, restraining tumour burden. Finally, recombinant IL-9 injection enhances tumour control in dysbiotic mice. Host-microbiota interactions are required for adequate differentiation and antitumour function of IL-9-producing T cells.