Merck
Pitolisant and intravenous cocaine self-administration in mice.
European journal of pharmacology (2019-02-17)
Benjamin Huyts, Christian Brabant, Ezio Tirelli
ABSTRACT
Pitolisant, a selective inverse agonist for the histamine H3 receptor, is a new treatment for adults suffering from narcolepsy. Numerous studies have shown that striatal H3 receptors can modulate the activity of the dopamine mesolimbic system, a neuronal pathway that plays a crucial role in drug addiction. Therefore, it is important to guarantee that pitolisant has no abuse potential and does not potentiate the behavioral effects of psychostimulants. The present study tested the effects of pitolisant on cocaine reinforcement in C57BL/6J mice using the intravenous self-administration technique. Mice were trained to self-administer cocaine intravenously. After the acquisition of cocaine self-administration, pitolisant was tested on cocaine self-administration under different schedules of reinforcement (fixed ratio and progressive ratio). In another group of mice, cocaine was replaced with pitolisant after the acquisition of cocaine self-administration. Finally, a group of mice was trained to self-administer pitolisant intravenously and directly compared to mice trained to self-administer cocaine under the same conditions. Our results indicate that pitolisant does not influence the reinforcing effects of cocaine under any of the experimental conditions used in this study. Moreover, pitolisant has no reinforcing properties alone when tested in the self-administration paradigm. Our results offer more evidence to support the hypothesis that pitolisant is not addictive. In addition, pitolisant does not alter the reinforcing effects of cocaine. Finally, the present study provides no evidence for a significant involvement of histamine H3 receptors in cocaine dependence.
Sigma-Aldrich
Pitolisant hydrochloride, ≥98% (HPLC)