Merck
3' Uridylation Confers miRNAs with Non-canonical Target Repertoires.
Molecular cell (2019-06-11)
Acong Yang, Xavier Bofill-De Ros, Tie-Juan Shao, Minjie Jiang, Katherine Li, Patricia Villanueva, Lisheng Dai, Shuo Gu
ABSTRACT
Many microRNAs (miRNAs) exist alongside abundant miRNA isoforms (isomiRs), most of which arise from post-maturation sequence modifications such as 3' uridylation. However, the ways in which these sequence modifications affect miRNA function remain poorly understood. Here, using human miR-27a in cell lines as a model, we discovered that a nonfunctional target site unable to base-pair extensively with the miRNA seed sequence can regain function when an upstream adenosine is able to base-pair with a post-transcriptionally added uridine in the miR-27a tail. This tail-U-mediated repression (TUMR) is abolished in cells lacking the uridylation enzymes TUT4 and TUT7, indicating that uridylation alters miRNA function by modulating target recognition. We identified a set of non-canonical targets in human cells that are specifically regulated by uridylated miR-27a. We provide evidence that TUMR expands the targets of other endogenous miRNAs. Our study reveals a function of uridylated isomiRs in regulating non-canonical miRNA targets.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture