Merck
  • Lack of a Pharmacokinetic Interaction Between SYM-1219 Granules Containing 2 Grams of Secnidazole and a Combined Oral Contraceptive in a Phase 1, Randomized, Open-Label Study in Healthy Female Volunteers.
Lack of a Pharmacokinetic Interaction Between SYM-1219 Granules Containing 2 Grams of Secnidazole and a Combined Oral Contraceptive in a Phase 1, Randomized, Open-Label Study in Healthy Female Volunteers.
Advances in therapy (2016-10-17)
Helen S Pentikis, Nikki Adetoro, Carol J Braun
ABSTRACT
Bacterial vaginosis (BV) is a serious infection that is the most common vaginal infection in women of childbearing potential. SYM-1219 is a novel, granule formulation containing 2 g of secnidazole that is being developed as a single, oral dose to treat women with BV. Because many of the women diagnosed with BV use hormonal contraception, the effect of SYM-1219 on the pharmacokinetics (PK) of commonly prescribed oral contraceptive drugs, ethinyl estradiol (EE2), and norethindrone (NET) was evaluated. This two-period, randomized, open-label study examined effects in 54 healthy female subjects. During the first period of the study, each subject received EE2 0.035-mg/NET 1-mg tablets. During the second period of the study, subjects were randomized to receive either EE2 0.035-mg/NET 1-mg tablets with concomitant 2-g SYM-1219 or 2-g SYM-1219 followed by EE2 0.035-mg/NET 1-mg tablets 1 day later. The PK of EE2 and NET were analyzed for 24 h following administration. Coadministration of SYM-1219 and EE2/NET, either on the same day or 1 day apart, had no clinically relevant effects on the bioavailability of EE2 or NET. The combined use of SYM-1219 with EE2/NET was well tolerated. Taken together, these results indicate that contraceptive efficacy should be maintained during coadministration of SYM-1219 and EE2/NET. SYM-1219 is a valuable single-dose treatment option for women with BV that will not interfere with combined oral contraceptive methods. Symbiomix Therapeutics.
Sigma-Aldrich
Secnidazole, ≥98% (HPLC)