Merck
  • Approaching a complete repository of sequence-verified protein-encoding clones for Saccharomyces cerevisiae.
Approaching a complete repository of sequence-verified protein-encoding clones for Saccharomyces cerevisiae.
Genome research (2007-02-27)
Yanhui Hu, Andreas Rolfs, Bhupinder Bhullar, Tellamraju V S Murthy, Cong Zhu, Michael F Berger, Anamaria A Camargo, Fontina Kelley, Seamus McCarron, Daniel Jepson, Aaron Richardson, Jacob Raphael, Donna Moreira, Elena Taycher, Dongmei Zuo, Stephanie Mohr, Michael F Kane, Janice Williamson, Andrew Simpson, Martha L Bulyk, Edward Harlow, Gerald Marsischky, Richard D Kolodner, Joshua LaBaer
ABSTRACT
The availability of an annotated genome sequence for the yeast Saccharomyces cerevisiae has made possible the proteome-scale study of protein function and protein-protein interactions. These studies rely on availability of cloned open reading frame (ORF) collections that can be used for cell-free or cell-based protein expression. Several yeast ORF collections are available, but their use and data interpretation can be hindered by reliance on now out-of-date annotations, the inflexible presence of N- or C-terminal tags, and/or the unknown presence of mutations introduced during the cloning process. High-throughput biochemical and genetic analyses would benefit from a "gold standard" (fully sequence-verified, high-quality) ORF collection, which allows for high confidence in and reproducibility of experimental results. Here, we describe Yeast FLEXGene, a S. cerevisiae protein-coding clone collection that covers over 5000 predicted protein-coding sequences. The clone set covers 87% of the current S. cerevisiae genome annotation and includes full sequencing of each ORF insert. Availability of this collection makes possible a wide variety of studies from purified proteins to mutation suppression analysis, which should contribute to a global understanding of yeast protein function.