Merck
  • Caspase-11 cleaves gasdermin D for non-canonical inflammasome signalling.
Caspase-11 cleaves gasdermin D for non-canonical inflammasome signalling.
Nature (2015-09-17)
Nobuhiko Kayagaki, Irma B Stowe, Bettina L Lee, Karen O'Rourke, Keith Anderson, Søren Warming, Trinna Cuellar, Benjamin Haley, Merone Roose-Girma, Qui T Phung, Peter S Liu, Jennie R Lill, Hong Li, Jiansheng Wu, Sarah Kummerfeld, Juan Zhang, Wyne P Lee, Scott J Snipas, Guy S Salvesen, Lucy X Morris, Linda Fitzgerald, Yafei Zhang, Edward M Bertram, Christopher C Goodnow, Vishva M Dixit
ABSTRACT
Intracellular lipopolysaccharide from Gram-negative bacteria including Escherichia coli, Salmonella typhimurium, Shigella flexneri, and Burkholderia thailandensis activates mouse caspase-11, causing pyroptotic cell death, interleukin-1β processing, and lethal septic shock. How caspase-11 executes these downstream signalling events is largely unknown. Here we show that gasdermin D is essential for caspase-11-dependent pyroptosis and interleukin-1β maturation. A forward genetic screen with ethyl-N-nitrosourea-mutagenized mice links Gsdmd to the intracellular lipopolysaccharide response. Macrophages from Gsdmd(-/-) mice generated by gene targeting also exhibit defective pyroptosis and interleukin-1β secretion induced by cytoplasmic lipopolysaccharide or Gram-negative bacteria. In addition, Gsdmd(-/-) mice are protected from a lethal dose of lipopolysaccharide. Mechanistically, caspase-11 cleaves gasdermin D, and the resulting amino-terminal fragment promotes both pyroptosis and NLRP3-dependent activation of caspase-1 in a cell-intrinsic manner. Our data identify gasdermin D as a critical target of caspase-11 and a key mediator of the host response against Gram-negative bacteria.