Skip to Content
Merck
Tropisetron impurity B European Pharmacopoeia (EP) Reference Standard

Tropisetron impurity B

Tropisetron impurity B European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Indole-3-carboxylic acid, β-Indolylcarboxylic acid, 3-Carboxyindole, 3-Indole formic acid, 3-Indolylcarboxylic acid, Indole-β-carboxylic acid
Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing