Skip to Content
Merck
Terpineol mixture of isomers, analytical standard

Terpineol

Terpineol mixture of isomers, analytical standard
Empirical Formula (Hill Notation):
C10H18O
Molecular Weight:
154.25
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-4 of 4