Skip to Content
Merck
Trapidil impurity B European Pharmacopoeia (EP) Reference Standard

Amitrol

Trapidil impurity B European Pharmacopoeia (EP) Reference Standard
Synonym(s):
3-Amino-1,2,4-triazole, 1,2,4-Triazol-3-amine, 3-AT, Amitrol
Empirical Formula (Hill Notation):
C2H4N4
CAS Number:
Molecular Weight:
84.08

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing