Skip to Content
Merck
vibrant-m

Salmeterol Xinafoate

vibrant-m
Synonym(s):
Salmeterol xinafoate, (±) 4-Hydroxy-a1-[[[6-(4-phenylbutoxy)hexyl]amino]m-ethyl]-1,3-benzenedimethanol xinafoate, GR 33343X xinafoate
Empirical Formula (Hill Notation):
C25H37NO4 · C11H8O3
CAS Number:
Molecular Weight:
603.75

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique