Skip to Content
Merck
Trehalose dihydrate European Pharmacopoeia (EP) Reference Standard

Trehalose

Trehalose dihydrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
D-(+)-Trehalose dihydrate, α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside
Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33

Products

Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique