Skip to Content
Merck
vibrant-m

Magnesium Standard for AAS

vibrant-m
Synonym(s):
Magnesium nitrate hexahydrate
Linear Formula:
Mg(NO3)2 · 6 H2O
CAS Number:
Molecular Weight:
256.41

Products

Shipping
Brand
Formula Weight
Product Line
Format
Quality Segment
Special Grade
Technique