Skip to Content
Merck
Acetaminophen Related Compound J United States Pharmacopeia (USP) Reference Standard

Acetaminophen Related Compound J

Acetaminophen Related Compound J United States Pharmacopeia (USP) Reference Standard
Synonym(s):
4′-Chloroacetanilide, N-(4-Chlorophenyl)acetamide, Acetic acid 4-chloroanilide, NSC 40563, NSC 444
Linear Formula:
CH3CONHC6H4Cl
CAS Number:
Molecular Weight:
169.61

Products

Shipping
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique