Skip to Content
DE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium acetate solution
Linear Formula:
CH
3
COOK
CAS Number:
127-08-2
Molecular Weight:
98.14
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (2)
Formula Weight
pH Value
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
95843
Potassium acetate solution,
BioUltra, for molecular biology, 5 M in H
2
O
Expand
View Pricing
SAE0157
Potassium acetate solution,
100 mM, pH 5.0, 0.2 μM filtered
Expand
View Pricing