DCC
Synonym(s):
N,N′-Dicyclohexylcarbodiimide solution
Linear Formula:
C6H11N=C=NC6H11
CAS Number:
Molecular Weight:
206.33
Products
Shipping
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing