Skip to Content
Merck
Peptone Water, phosphate-buffered suitable for microbiology, NutriSelect® Plus

Peptone Water, phosphate-buffered

Peptone Water, phosphate-buffered suitable for microbiology, NutriSelect® Plus
Synonym(s):
Buffered Peptone Water, Tryptone Phosphate Water

Products

Shipping
Product Category
Brand
pH Value
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing