Skip to Content
Merck
(+)-α-Pinene analytical standard

(+)-α-Pinene

(+)-α-Pinene analytical standard
Synonym(s):
(1R,5R)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene, (1R,5R)-2-Pinene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing