![Carvacrol methyl ether phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/110/097/5af597d1-5256-4cf3-845e-d61266a12e82/640/5af597d1-5256-4cf3-845e-d61266a12e82.png)
Carvacrol methyl ether
![Carvacrol methyl ether phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/110/097/5af597d1-5256-4cf3-845e-d61266a12e82/640/5af597d1-5256-4cf3-845e-d61266a12e82.png)
Synonym(s):
2-Methoxy-p-cymol, 4-Isopropyl-2-methoxytoluene, 5-Isopropyl-2-methylanisole, Methylcarvacrol
Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Products
Shipping
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing