![Benzonase® purity grade II (>90%) for biotechnology](/deepweb/assets/sigmaaldrich/product/images/279/686/79b39ae3-14df-4933-a710-4a21092dc59c/640/79b39ae3-14df-4933-a710-4a21092dc59c.jpg)
Benzonase® purity grade II (>90%) for biotechnology
![Benzonase® purity grade II (>90%) for biotechnology](/deepweb/assets/sigmaaldrich/product/images/279/686/79b39ae3-14df-4933-a710-4a21092dc59c/640/79b39ae3-14df-4933-a710-4a21092dc59c.jpg)
Synonym(s):
Endonuclease from Serratia marcescens
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Recombinant Host
Quality Segment
Application
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing