Skip to Content
Merck
vibrant-m

AmberChrom 50WX4

vibrant-m
CAS Number:

Products

Product Category
Brand
Physical Form
Separation Technique
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing