Skip to Content
Merck
dTTP Lithium salt

Thymidine 5′-triphosphate sodium salt

dTTP Lithium salt
Synonym(s):
TTP, dTTP, Thymidine 5′-triphosphate sodium salt
Empirical Formula (Hill Notation):
C10H16N2O14P3Na
CAS Number:
Molecular Weight:
504.15

Products

Brand
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2