Skip to Content
Merck
Lithium chloride AnhydroBeads™, −10 mesh, ≥99.9% trace metals basis

Lithium chloride

Lithium chloride AnhydroBeads™, −10 mesh, ≥99.9% trace metals basis
Synonym(s):
Lithium chloride
Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:

Products

Shipping
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade