Skip to Content
Merck
Ethylenediaminetetraacetic acid disodium salt dihydrate ≥97.0% (KT)

Ethylenediaminetetraacetic acid disodium salt dihydrate

Ethylenediaminetetraacetic acid disodium salt dihydrate ≥97.0% (KT)
Synonym(s):
Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2, Ethylenediaminetetraacetic acid disodium salt dihydrate
Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Reaction Type
Reagent Type
API Family
Reference Material Type
Special Grade
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing