Skip to Content
Merck
Fluconazole ≥98% (HPLC), powder

Fluconazole

Fluconazole ≥98% (HPLC), powder
Synonym(s):
2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27

Products

Shipping
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing