Skip to Content
Merck
Parteck® SI 450 (Sorbitol) EMPROVE® ESSENTIAL, NF, FCC, JSFA

D-Sorbitol

Parteck® SI 450 (Sorbitol) EMPROVE® ESSENTIAL, NF, FCC, JSFA
Synonym(s):
D-Glucitol, Parteck® Sorbitol, D-Sorbitol
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17

Products

Shipping
Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Special Grade
Technique