Skip to Content
Merck
TWEEN® 20 Vetec™, reagent grade, 40%

TWEEN® 20

TWEEN® 20 Vetec™, reagent grade, 40%
Synonym(s):
Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, TWEEN® 20, Polysorbate 20
CAS Number:

Products

Shipping
Product Category
Brand
Sustainability
Biological Source
Color
Feature
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Special Grade
Technique