Skip to Content
DE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polyethylenimine, branched
Synonym(s):
PEI
Linear Formula:
(CH
2
CH
2
NH)
n
CAS Number:
9002-98-6
Products
Shipping
Ships Today (1)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
408727
Polyethylenimine, branched,
average M
w
~25,000 by LS, average M
n
~10,000 by GPC, branched
Expand
View Pricing
765090
Polyethylenimine, branched,
average M
n
10,000, PDI ≤1.3
Expand
View Pricing
764604
Polyethylenimine, branched,
average M
n
2100, PDI <1.3
Expand
View Pricing
764582
Polyethylenimine, branched,
average M
n
5,000, PDI ≤1.3
Expand
View Pricing