Product Name
Trimethyloctadecylammonium chloride ≥95.0% (calc. on dry substance, T)
Product Number
53112
Product Brand
SIGMA
CAS Number
112-03-8
Molecular Weight
348.05
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder or Crystals |
ARGENTOMETR. TITRATION | 95.0 - 105.0 % |
SOLUBILITY (COLOR) | Colorless |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 5MG IN 10ML WATER |
PH | 5.5 - 8.5 |
WATER | ≤ 3.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
UV Absorbance | UV at 225 nm ≤ 0.080 |
UV Absorbance | UV at 260 nm ≤ 0.060 |
UV Absorbance | UV at 280 nm ≤ 0.040 |
UV Absorbance | UV at 340 nm ≤ 0.020 |