CWA-1171
Anti-Human LIF-R (1C7) ColorWheel® Dye-Ready mAb
Synonym(s):
Leukemia inhibitory factor receptor;LIF receptor;LIF-R; D-factor/LIF receptor;CD118
About This Item
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
1C7, monoclonal
product line
ColorWheel®
form
lyophilized
purified by
using protein G
species reactivity
human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
flow cytometry: suitable
isotype
IgG1κ
epitope sequence
Extracellular domain
Protein ID accession no.
UniProt accession no.
compatibility
for use with ColorWheel® Dyes (Required, (Sold Separately))
greener alternative category
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... Lifr(3977)
General description
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in CHO cells stably transfected with D1IgD2-mycDAF.
Flow Cytometry Analysis: Staining of one million Staining of one million CHO cells stably transfected with D1IgD2-mycDAF was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1171, Anti-Human LIF-R (1C7) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.
Tested Applications
Flow Cytometry Analysis: Staining of one million Staining of one million CHO cells stably transfected with D1IgD2-mycDAF was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1171, Anti-Human LIF-R (1C7) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or a 1:1 mixture of an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Compatibility
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service