Skip to Content
Merck
All Photos(1)

Key Documents

S4181

Millipore

Sabouraud maltose agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shelf life

limited shelf life, expiry date on the label

Quality Level

pH

5.6±0.2 (25 °C)

application(s)

microbiology

suitability

selective by low pH for Aspergillus spp.
selective by low pH for Candida spp.
selective by low pH for Penicillium spp.
selective by low pH for Pichia spp.
selective by low pH for Saccharomyces spp.
selective by low pH for Zygosaccharomyces spp.
selective by low pH for molds (General Media)
selective by low pH for yeasts (General Media)

Application

Used as an excellent medium for the propagation of molds and yeasts, particularly parasitic fungi concerned with the skin and scalp.

Components

Components (g/L)
Mycological Peptone 10.00
Maltose 40.00
Agar 15.00

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabouraud, K.
Ann. Dermatol. Syphilol., 3, 1061-1061 (1892)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service