Skip to Content
Merck
All Photos(1)

Key Documents

06203-M

Sigma-Aldrich

Sodium hydroxide

puriss., meets analytical specification of Ph. Eur., BP, NF, E524, ≥98.0%, pellets

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

grade

puriss.

Assay

≥98.0%

form

pellets

quality

meets analytical specification of Ph. Eur., BP, NF, E524

impurities

residual solvents, complies
≤0.0005% total nitrogen (N)
≤0.5% Na2CO3
≤20 ppm heavy metals (as Pb)

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

anion traces

chloride (Cl-): ≤20 ppm
phosphate (PO43-): ≤0.001%
silicate (as SiO2): ≤0.01%
sulfate (SO42-): ≤30 ppm

cation traces

Al: ≤0.001%
As: ≤2 ppm
Ca: ≤0.001%
Fe: ≤10 ppm
Hg: ≤1 ppm
K: ≤0.1%
Pb: ≤0.5 ppm

suitability

complies for appearance of solution

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Important notice
This product will be phased out towards early 2019. There is a replacement product available : 06203

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Roberts et al.
Analytical chemistry, 69(11), 2035-2042 (1997-06-01)
This report describes a UV laser photoablation method for the production of miniaturized liquid-handling systems on polymer substrate chips. The fabrication of fluid channel and reservoir networks is accomplished by firing 200 mJ pulses from an UV excimer laser at
Chemoselective, solvent-free aldol condensation reaction.
Raston CL and Scott JL.
Green Chemistry, 2(2), 49-52 (2000)
M Chevallet et al.
Electrophoresis, 19(11), 1901-1909 (1998-09-18)
Severe quantitative loss of protein is often observed in high-resolution two-dimensional electrophoresis of membrane proteins, while the resolution is usually not affected. To improve the solubility of proteins in this technique, we tested denaturing cocktails containing various detergents and chaotropes.
Kinetics of sodium borohydride direct oxidation and oxygen reduction in sodium hydroxide electrolyte: Part I. BH4- electro-oxidation on Au and Ag catalysts.
Chatenet M, et al.
Electrochimica Acta, 51(25), 5459-5467 (2006)
Silvio Carrettin et al.
Chemical communications (Cambridge, England), (7)(7), 696-697 (2002-07-18)
Glycerol is oxidised to glyceric acid with 100% selectivity using either 1% Au/charcoal or 1% Au/graphite catalyst under mild reaction conditions (60 degrees C, 3 h, water as solvent).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service