263044
Erbium
powder, −40 mesh, 99.9% trace rare earth metals basis
About This Item
Recommended Products
Assay
99.9% trace rare earth metals basis
form
powder
reaction suitability
reagent type: catalyst
core: erbium
resistivity
86 μΩ-cm, 20°C
particle size
−40 mesh
bp
2868 °C (lit.)
mp
1529 °C (lit.)
density
9.062 g/mL at 25 °C (lit.)
SMILES string
[Er]
InChI
1S/Er
InChI key
UYAHIZSMUZPPFV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- The role of Cerium, Europium and Erbium doped TiO2 photocatalysts in water treatment: A mini-review: This review explores the application of Erbium-doped TiO2 in water treatment for removing contaminants (Cerrato et al., 2022).
- Quantum storage of entangled telecom-wavelength photons in an erbium-doped optical fibre: The study demonstrates quantum storage capabilities using erbium-doped optical fibers for telecommunications (Saglamyurek et al., 2015).
- Twenty-three–millisecond electron spin coherence of erbium ions in a natural-abundance crystal: This paper discusses the long electron spin coherence times of erbium ions in crystals, relevant for quantum computing applications (Le Dantec et al., 2021).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.
Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.
Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service