RPMI-1640 Media Formulation
Component | R0883 [1x] g/L | R1145 [10x] g/L | R1383 g/L | R1780 [1x] g/L | R2405 [1x] g/L | R6504 g/L | R8758 [1x] g/L |
---|---|---|---|---|---|---|---|
Inorganic Salts | |||||||
Calcium Nitrate • 4H2O | 0.1 | 1 | 0.1 | 0.1 | 0.1 | 0.1 | 0.1 |
Magnesium Sulfate (anhydrous) | 0.04884 | 0.4884 | 0.04884 | 0.04884 | 0.04884 | 0.04884 | 0.04884 |
Potassium Chloride | 0.4 | 4 | 0.4 | 0.4 | 0.4 | 0.4 | 0.4 |
Sodium Bicarbonate | 2 | — | — | 2 | 2 | — | 2 |
Sodium Chloride | 6 | 60 | 6 | 6 | 6 | 6 | 6 |
Sodium Phosphate Dibasic (anhydrous) | 0.8 | 8 | 0.8 | 0.8 | 0.8 | 0.8 | 0.8 |
Amino Acids | |||||||
L-Alanyl-L-Glutamine | — | — | — | — | 0.4344 | — | — |
L-Arginine | 0.2 | 2 | 0.2 | — | 0.2 | 0.2 | 0.2 |
L-Asparagine (anhydrous) | 0.05 | 0.5 | 0.05 | 0.05 | 0.05 | 0.05 | 0.05 |
L-Aspartic Acid | 0.02 | 0.2 | 0.02 | 0.02 | 0.02 | 0.02 | 0.02 |
L-Cystine • 2HCl | 0.0652 | 0.652 | 0.0652 | 0.0652 | 0.0652 | 0.0652 | 0.0652 |
L-Glutamic Acid | 0.02 | 0.2 | 0.02 | 0.02 | 0.02 | 0.02 | 0.02 |
L-Glutamine | — | — | 0.3 | 0.3 | — | 0.3 | 0.3 |
Glycine | 0.01 | 0.1 | 0.01 | 0.01 | 0.01 | 0.01 | 0.01 |
L-Histidine | 0.015 | 0.15 | 0.015 | 0.015 | 0.015 | 0.015 | 0.015 |
Hydroxy-L-Proline | 0.02 | 0.2 | 0.02 | 0.02 | 0.02 | 0.02 | 0.02 |
L-Isoleucine | 0.05 | 0.5 | 0.05 | 0.05 | 0.05 | 0.05 | 0.05 |
L-Leucine | 0.05 | 0.5 | 0.05 | — | 0.05 | 0.05 | 0.05 |
L-Lysine • HCl | 0.04 | 0.4 | 0.04 | — | 0.04 | 0.04 | 0.04 |
L-Methionine | 0.015 | 0.15 | 0.015 | 0.015 | 0.015 | 0.015 | 0.015 |
L-Phenylalanine | 0.015 | 0.15 | 0.015 | 0.015 | 0.015 | 0.015 | 0.015 |
L-Proline | 0.02 | 0.2 | 0.02 | 0.02 | 0.02 | 0.02 | 0.02 |
L-Serine | 0.03 | 0.3 | 0.03 | 0.03 | 0.03 | 0.03 | 0.03 |
L-Threonine | 0.02 | 0.2 | 0.02 | 0.02 | 0.02 | 0.02 | 0.02 |
L-Tryptophan | 0.005 | 0.05 | 0.005 | 0.005 | 0.005 | 0.005 | 0.005 |
L-Tyrosine • 2Na • 2H2O | 0.02883 | 0.2883 | 0.02883 | 0.02883 | 0.02883 | 0.02883 | 0.02883 |
L-Valine | 0.02 | 0.2 | 0.02 | 0.02 | 0.02 | 0.02 | 0.02 |
Vitamins | |||||||
D-Biotin | 0.0002 | 0.002 | 0.0002 | 0.002 | 0.002 | 0.0002 | 0.0002 |
Choline Chloride | 0.003 | 0.03 | 0.003 | 0.003 | 0.003 | 0.003 | 0.003 |
Folic Acid | 0.001 | — | 0.001 | 0.001 | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.035 | 0.35 | 0.035 | 0.035 | 0.035 | 0.035 | 0.035 |
Niacinamide | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 | 0.001 | 0.001 |
p-Aminobenzoic Acid | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.00025 | 0.0025 | 0.00025 | 0.00025 | 0.00025 | 0.00025 | 0.00025 |
Pyridoxine • HCl | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 | 0.001 | 0.001 |
Riboflavin | 0.0002 | 0.002 | 0.0002 | 0.0002 | 0.0002 | 0.0002 | 0.0002 |
Thiamine • HCl | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 | 0.001 | 0.001 |
Vitamin B12 | 0.000005 | 0.00005 | 0.000005 | 0.000005 | 0.000005 | 0.000005 | 0.000005 |
Other | |||||||
D-Glucose | 2 | 20 | — | 2 | 2 | 2 | 2 |
Glutathione (reduced) | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 | 0.001 | 0.001 |
Phenol Red • Na | 0.0053 | 0.053 | 0.0053 | — | 0.0053 | 0.0053 | 0.0053 |
Add | |||||||
L-Glutamine | 0.3 | 0.3 at 1× | — | — | — | — | — |
Sodium Bicarbonate | — | 2.0 at 1× | 2 | — | — | 2 | — |
Component | R7638 [1x] g/L |
---|---|
Inorganic Salts | |
Calcium Nitrate • 4H2O | 0.1 |
Magnesium Sulfate (anhydrous) | 0.04884 |
Potassium Chloride | 0.4 |
Sodium Bicarbonate | 1 |
Sodium Chloride | 6.4 |
Sodium Phosphate Dibasic (anhydrous) | 0.8 |
Amino Acids | |
L-Arginine | 0.2 |
L-Asparagine (anhydrous) | 0.05 |
L-Aspartic Acid | 0.02 |
L-Cystine • 2HCl | 0.0652 |
L-Glutamic Acid | 0.02 |
Glycine | 0.01 |
L-Histidine | 0.015 |
Hydroxy-L-Proline | 0.02 |
L-Isoleucine | 0.05 |
L-Leucine | 0.05 |
L-Lysine • HCl | 0.04 |
L-Methionine | 0.015 |
L-Phenylalanine | 0.015 |
L-Proline | 0.02 |
L-Serine | 0.03 |
L-Threonine | 0.02 |
L-Tryptophan | 0.005 |
L-Tyrosine • 2Na • 2H2O | 0.02883 |
L-Valine | 0.02 |
Vitamins | |
D-Biotin | 0.0002 |
Choline Chloride | 0.003 |
Folic Acid | 0.001 |
myo-Inositol | 0.035 |
Niacinamide | 0.001 |
p-Aminobenzoic Acid | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.00025 |
Pyridoxine • HCl | 0.001 |
Riboflavin | 0.0002 |
Thiamine • HCl | 0.001 |
Vitamin B12 | 0.000005 |
Other | |
D-Glucose | 2 |
Glutathione (reduced) | 0.001 |
HEPES | 4.77 |
Phenol Red • Na | 0.0053 |
Add | |
L-Glutamine | 0.3 |
Component | R4130 g/L | R5886 [1x] g/L | R8005 g/L |
---|---|---|---|
Inorganic Salts | |||
Calcium Nitrate • 4H2O | 0.1 | 0.1 | 0.1 |
Magnesium Sulfate (anhydrous) | 0.04884 | 0.04884 | 0.04884 |
Potassium Chloride | 0.4 | 0.4 | 0.4 |
Sodium Bicarbonate | — | 2 | — |
Sodium Chloride | 6 | 6 | 5.9 |
Sodium Phosphate Dibasic (anhydrous) | 0.8 | 0.8 | 0.8 |
Amino Acids | |||
L-Arginine | 0.2 | 0.2 | 0.2 |
L-Asparagine (anhydrous) | 0.05 | 0.05 | 0.05 |
L-Aspartic Acid | 0.02 | 0.02 | 0.02 |
L-Cystine • 2HCl | 0.0652 | 0.0652 | 0.0652 |
L-Glutamic Acid | 0.02 | 0.02 | 0.02 |
L-Glutamine | 0.3 | — | 0.3 |
Glycine | 0.01 | 0.01 | 0.01 |
L-Histidine | 0.015 | 0.015 | 0.015 |
Hydroxy-L-Proline | 0.02 | 0.02 | 0.02 |
L-Isoleucine | 0.05 | 0.05 | 0.05 |
L-Leucine | 0.05 | 0.05 | 0.05 |
L-Lysine • HCl | 0.04 | 0.04 | 0.04 |
L-Methionine | 0.015 | 0.015 | 0.015 |
L-Phenylalanine | 0.015 | 0.015 | 0.015 |
L-Proline | 0.02 | 0.02 | 0.02 |
L-Serine | 0.03 | 0.03 | 0.03 |
L-Threonine | 0.02 | 0.02 | 0.02 |
L-Tryptophan | 0.005 | 0.005 | 0.005 |
L-Tyrosine • 2Na • 2H2O | 0.02883 | 0.02883 | 0.02883 |
L-Valine | 0.02 | 0.02 | 0.02 |
Vitamins | |||
D-Biotin | 0.0002 | 0.0002 | 0.0002 |
Choline Chloride | 0.003 | 0.003 | 0.003 |
Folic Acid | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.035 | 0.035 | 0.035 |
Niacinamide | 0.001 | 0.001 | 0.001 |
p-Aminobenzoic Acid | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.00025 | 0.00025 | 0.00025 |
Pyridoxine • HCl | 0.001 | 0.001 | 0.001 |
Riboflavin | 0.0002 | 0.0002 | 0.0002 |
Thiamine • HCl | 0.001 | 0.001 | 0.001 |
Vitamin B12 | 0.000005 | 0.000005 | 0.000005 |
Other | |||
D-Glucose | 2 | 2 | 4.5 |
Glutathione (reduced) | 0.001 | 0.001 | 0.001 |
HEPES | 5.96 | 5.96 | 3.5745 |
Phenol Red • Na | 0.0053 | 0.0053 | 0.0053 |
Add | |||
L-Glutamine | — | 0.3 | — |
Sodium Bicarbonate | 2 | — | 2 |
Component | R7388 [1x] g/L | R7509 [1x] g/L | R7513 [1x] g/L | R8755 g/L |
---|---|---|---|---|
Inorganic Salts | ||||
Calcium Nitrate • 4H2O | 0.1 | 0.1 | 0.1 | 0.1 |
Magnesium Sulfate (anhydrous) | 0.04884 | 0.04884 | 0.04884 | 0.04884 |
Potassium Chloride | 0.4 | 0.4 | 0.4 | 0.4 |
Sodium Bicarbonate | — | 2 | 2 | — |
Sodium Chloride | 6 | 6 | 6 | 6 |
Sodium Phosphate Dibasic (anhydrous) | 0.8 | 0.8 | 0.8 | 0.8 |
Amino Acids | ||||
L-Arginine | 0.2 | 0.2 | 0.2 | 0.2 |
L-Asparagine (anhydrous) | 0.05 | 0.05 | 0.05 | 0.05 |
L-Aspartic Acid | 0.02 | 0.02 | 0.02 | 0.02 |
L-Cystine • 2HCl | 0.0652 | 0.0652 | — | 0.0652 |
L-Glutamic Acid | 0.02 | 0.02 | 0.02 | 0.02 |
L-Glutamine | 0.3 | — | — | 0.3 |
Glycine | 0.01 | 0.01 | 0.01 | 0.01 |
L-Histidine | 0.015 | 0.015 | 0.015 | 0.015 |
Hydroxy-L-Proline | 0.02 | 0.02 | 0.02 | 0.02 |
L-Isoleucine | 0.05 | 0.05 | 0.05 | 0.05 |
L-Leucine | 0.05 | 0.05 | 0.05 | 0.05 |
L-Lysine • HCl | 0.04 | 0.04 | 0.04 | 0.04 |
L-Methionine | 0.015 | 0.015 | — | 0.015 |
L-Phenylalanine | 0.015 | 0.015 | 0.015 | 0.015 |
L-Proline | 0.02 | 0.02 | 0.02 | 0.02 |
L-Serine | 0.03 | 0.03 | 0.03 | 0.03 |
L-Threonine | 0.02 | 0.02 | 0.02 | 0.02 |
L-Tryptophan | 0.005 | 0.005 | 0.005 | 0.005 |
L-Tyrosine • 2Na • 2H2O | 0.02883 | 0.02883 | 0.02883 | 0.02883 |
L-Valine | 0.02 | 0.02 | 0.02 | 0.02 |
Vitamins | ||||
D-Biotin | 0.0002 | 0.0002 | 0.0002 | 0.0002 |
Choline Chloride | 0.003 | 0.003 | 0.003 | 0.003 |
Folic Acid | 0.001 | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.035 | 0.035 | 0.035 | 0.035 |
Niacinamide | 0.001 | 0.001 | 0.001 | 0.001 |
p-Aminobenzoic Acid | 0.001 | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.00025 | 0.00025 | 0.00025 | 0.00025 |
Pyridoxine • HCl | 0.001 | 0.001 | 0.001 | 0.001 |
Riboflavin | 0.0002 | 0.0002 | 0.0002 | 0.0002 |
Thiamine • HCl | 0.001 | 0.001 | 0.001 | 0.001 |
Vitamin B12 | 0.000005 | 0.000005 | 0.000005 | 0.000005 |
Other | ||||
D-Glucose | 2 | 2 | 2 | 2 |
Glutathione (reduced) | 0.001 | 0.001 | 0.001 | 0.001 |
HEPES | 4.77 | — | — | — |
Phenol Red • Na | 0.0053 | — | 0.0053 | — |
Add | ||||
L-Cystine • 2HCl | — | — | 0.0652 | — |
L-Glutamine | — | 0.3 | 0.3 | — |
L-Methionine | — | — | 0.015 | — |
Sodium Bicarbonate | 2 | — | — | 2 |
Component | R7755 g/L |
---|---|
Inorganic Salts | |
Calcium Nitrate • 4H2O | 0.1 |
Magnesium Sulfate (anhydrous) | 0.04884 |
Potassium Chloride | 0.4 |
Sodium Chloride | 6 |
Sodium Phosphate Dibasic (anhydrous) | 0.8 |
Succinic Acid • 6H2O • Na | 0.1 |
Succinic Acid (free acid) | 0.075 |
Amino Acids | |
L-Arginine | 0.2 |
L-Asparagine (anhydrous) | 0.05 |
L-Aspartic Acid | 0.02 |
L-Cystine • 2HCl | 0.0652 |
L-Glutamic Acid | 0.02 |
Glycine | 0.01 |
L-Histidine | 0.015 |
Hydroxy-L-Proline | 0.02 |
L-Isoleucine | 0.05 |
L-Leucine | 0.05 |
L-Lysine • HCl | 0.04 |
L-Methionine | 0.015 |
L-Phenylalanine | 0.015 |
L-Proline | 0.02 |
L-Serine | 0.03 |
L-Threonine | 0.02 |
L-Tryptophan | 0.005 |
L-Tyrosine | 0.02 |
L-Valine | 0.02 |
Vitamins | |
D-Biotin | 0.0002 |
Choline Bitartrate | 0.00544 |
Folic Acid | 0.001 |
myo-Inositol | 0.035 |
Niacinamide | 0.001 |
p-Aminobenzoic Acid | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.00025 |
Pyridoxine • HCl | 0.001 |
Riboflavin | 0.0002 |
Thiamine • HCl | 0.001 |
Vitamin B12 | 0.000005 |
Other | |
D-Glucose | 2 |
Glutathione (reduced) | 0.001 |
Phenol Red • Na | 0.00318 |
Add | |
L-Glutamine | 0.3 |
Sodium Bicarbonate | 2 |