Merck
GC Analysis of LSD (TMS Derivatives) on SLB®-5ms
GC Analysis of LSD (TMS Derivatives) on SLB®-5ms suitable for GC
Materialien
Analytische Säule
Produkt-Nr.
Beschreibung
Preisangaben
SLB®-5ms GC-Kapillarsäule
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
150 C (1.50 min), 20 C/min to 300 C (20 min)
inj. temp.
250 °C
MSD interface
330 °C
scan range
m/z 45-525
carrier gas
helium, 0.7 mL/min constant
injection
0.5 μL pulsed splitless, 30 psi. (0.20 min), purge on (1.5 min), purge flow (50.0 mL/min)
liner
2mm I.D., straight
sample
Derivatized LSD using BSTFA