HPLC Analysis of Dopamine and Related Compounds on Discovery® HS F5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery HS F5, 15 cm × 4.6 mm I.D., 5 μm particles (567516-U)
mobile phase
[A] 10 mM ammonium formate (pH 3.0 with concentrated formic acid), [B] acetonitrile
flow rate
1 mL/min, split to the MS
column temp.
35 °C
detector
MS in SIR (Selected Ion Recording) mode using (+) or (-) ESI mode
injection
25 μL
sample
50 μg/mL in 10 mM ammonium formate (pH 3.0 with formic acid)
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany