Merck
HPLC Analysis of t-BOC-Methionine Enantiomers on Astec® (R,R) P-CAP
HPLC Analysis of t-BOC-Methionine Enantiomers on Astec® (R,R) P-CAP™ application for HPLC
Materials
standard
Product No.
Description
Pricing
L-Methionine
from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
L-Methionine
reagent grade, ≥98% (HPLC)
DL-Methionine
BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
L-Methionine
Z-L-Methionine
97%
DL-Methionine
99%, FCC, FG
CONDITIONS
column
Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 μm particles (31024AST)
mobile phase
20 mM ammonium acetate in: (A) acetonitrile; (B) methanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
2 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC