Merck
  • Synthesis of graft copolymers based on hyaluronan and poly(3-hydroxyalkanoates).
Synthesis of graft copolymers based on hyaluronan and poly(3-hydroxyalkanoates).
Carbohydrate polymers (2017-06-06)
Gloria Huerta-Angeles, Martina Brandejsová, Rinat Nigmatullin, Kateřina Kopecká, Hana Vágnerová, Daniela Šmejkalová, Ipsita Roy, Vladimír Velebný
ABSTRACT
This work reports the synthesis and characterisation of new amphiphilic hyaluronan (HA) grafted with poly(3-hydroxyalkanoates) (PHAs) conjugates. Hydrolytic depolymerisation of PHAs was used for the synthesis of defined oligo(3-hydroxyalkanoates)-containing carboxylic terminal moieties. A kinetic study of the depolymerisation was followed to prepare oligomers of required molecular weight. PHAs were coupled with hydroxyl groups of HA mediated by N, N'-carbonyldiimidazole (CDI) or HSTU Tetramethyl-O-(N-succinimidyl) uronium hexafluorophosphate. For the first time, the covalent bonding of oligo derivatives of P(3-hydroxybutyrate), P(3-hydroxyoctanoate), P(3-hydroxyoctanoate-co-3-hydroxydecanoate) and P(3-hydroxyoctanoate-co-3-hydroxydecanoate-co-3-hydroxydodecanoate) and HA was achieved by "grafting to" strategy. Achieved grafting degree was a function of hydrophobicity of PHA, Mw and polarity of the solvent. The most suitable reaction conditions were observed for oligo (3-hydroxybutyrate) grafted to HA (grafting degree of 14%). Graft copolymers were characterized by FT-IR, NMR, DSC and SEC-MALLS. Graft copolymers can be physically loaded with hydrophobic drugs and may serve as drug delivery system.
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
N,N,N′,N′-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate, ≥99.0% (TLC/N)